accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5521-31-3

IUPAC Name: 7,18-dimethyl-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: pigment red 179
Molecular Weight (g/mol): 418.41
Molecular Formula: C26H14N2O4
InChi Key: PJQYNUFEEZFYIS-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=CC=C3C4=CC=C5C(=O)N(C)C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results