accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 5522-43-0

IUPAC Name: 1-nitropyrene
Synonyms: 1-nitropyrene
Molecular Weight (g/mol): 247.25
Molecular Formula: C16H9NO2
InChi Key: ALRLPDGCPYIVHP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results