accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH2NOOFFFNOO

CAS RN 55283-68-6

IUPAC Name: N-ethyl-N-(2-methylprop-2-en-1-yl)-2,6-dinitro-4-(trifluoromethyl)aniline
Synonyms: ethalfluralin
Molecular Weight (g/mol): 333.27
Molecular Formula: C13H14F3N3O4
InChi Key: PTFJIKYUEPWBMS-UHFFFAOYSA-N
SMILES: CCN(CC(C)=C)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results