accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CSNH3COOCH3H3CO

CAS RN 55285-14-8

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl N-[(dibutylamino)sulfanyl]-N-methylcarbamate
Synonyms: carbosulfan caswell no. 463C marshal dibutylaminosulfenylcarbofuran marshall posse epa pesticide chemical code 090602
Molecular Weight (g/mol): 380.55
Molecular Formula: C20H32N2O3S
InChi Key: JLQUFIHWVLZVTJ-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)SN(C)C(=O)OC1=CC=CC2=C1OC(C)(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results