accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 553-53-7

IUPAC Name: pyridine-3-carbohydrazide
Synonyms: pyridine-3-carbohydrazide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: KFUSANSHCADHNJ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CN=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)