accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 553-97-9

IUPAC Name: 2-methylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2-methyl-1,4-benzoquinone
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: VTWDKFNVVLAELH-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=CC1=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)