accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBr

CAS RN 55362-80-6

IUPAC Name: 9-bromononan-1-ol
Synonyms: 9-bromononan-1-ol
Molecular Weight (g/mol): 223.15
Molecular Formula: C9H19BrO
InChi Key: USJDOLXCPFASNV-UHFFFAOYSA-N
SMILES: OCCCCCCCCCBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)