accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3FFF(E/Z)

CAS RN 55364-28-8

IUPAC Name: trimethyl(3,3,3-trifluoroprop-1-en-1-yl)silane
Synonyms: trimethyl(3,3,3-trifluoroprop-1-en-1-yl)silane
Molecular Weight (g/mol): 168.23
Molecular Formula: C6H11F3Si
InChi Key: MBCUJGNIYCDYKD-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C=CC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results