accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 554-62-1

IUPAC Name: (2S,3S,4R)-2-aminooctadecane-1,3,4-triol
Synonyms: phytosphingosine (+)-D-ribo-phytosphingosine 8-(Z/E)-C18-phytosphingenine 8-(z/e)-c18-phytosphingenine C18-phytosphingosine 4D-hydroxysphinganine 4-D-hydroxysphinganine 4-hydroxysphinganine 4-D-hydroxy-sphinganine 4-d-hydroxysphinganine
Molecular Weight (g/mol): 317.51
Molecular Formula: C18H39NO3
InChi Key: AERBNCYCJBRYDG-KSZLIROESA-N
SMILES: CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@@H](N)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results