accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55401-12-2

IUPAC Name: λ³-nickel(3+) bis((1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide) tetrabutylazanium
Synonyms: λ³-nickel(3+) bis((1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide) tetrabutylammonium ion
Molecular Weight (g/mol): 581.52
Molecular Formula: C24H36N5NiS4
InChi Key: JETKTJCVEPVWKF-UHFFFAOYSA-J
SMILES: [Ni+3].[S-]C(C#N)=C([S-])C#N.[S-]C(C#N)=C([S-])C#N.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results