accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONCl

CAS RN 55403-91-3

IUPAC Name: 1-(2-chloro-4-nitrophenyl)pyrrolidine
Synonyms: 1-(2-chloro-4-nitrophenyl)pyrrolidine
Molecular Weight (g/mol): 226.66
Molecular Formula: C10H11ClN2O2
InChi Key: XMLGZIMCJDYHOD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCCC2)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results