accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 554405-17-3

ONNOOH

CAS RN 554405-17-3

IUPAC Name: 6-(morpholin-4-yl)pyridine-2-carboxylic acid
Synonyms: 6-(morpholin-4-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: SAZSDHZXRMXESN-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(=CC=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results