CAS RN 55464-99-8
CAS RN 55464-99-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amberlite™ IRP-69 ion-exchange resin, Thermo Scientific™
CAS: 55464-99-8 MDL Number: MFCD00132786