accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55488-51-2

IUPAC Name: decyl carbonochloridate
Synonyms: n-decyl chloroformate
Molecular Weight (g/mol): 220.74
Molecular Formula: C11H21ClO2
InChi Key: AZZCHVHSWUYCQA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC(Cl)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)