accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiO

CAS RN 555-24-8

IUPAC Name: lithium(1+) benzenolate
Synonyms: lithium(1+) phenolate
Molecular Weight (g/mol): 100.05
Molecular Formula: C6H5LiO
InChi Key: XAVQZBGEXVFCJI-UHFFFAOYSA-M
SMILES: [Li+].[O-]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results