CAS RN 555-37-3
IUPAC Name:
3-butyl-1-(3,4-dichlorophenyl)-3-methylurea
Synonyms:
neburon
Molecular Weight (g/mol):
275.17
Molecular Formula:
C12H16Cl2N2O
InChi Key:
CCGPUGMWYLICGL-UHFFFAOYSA-N
SMILES:
CCCCN(C)C(=O)NC1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Neburon, SPEX CertiPrep™
CAS: 555-37-3 Molecular Formula: C12H16Cl2N2O Molecular Weight (g/mol): 275.173 InChI Key: CCGPUGMWYLICGL-UHFFFAOYSA-N PubChem CID: 11145 ChEBI: CHEBI:7491 IUPAC Name: 1-butyl-3-(3,4-dichlorophenyl)-1-methylurea SMILES: CCCCN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl