accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 555-68-0

IUPAC Name: (2Z)-3-(3-nitrophenyl)prop-2-enoic acid
Synonyms: (2Z)-3-(3-nitrophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 193.16
Molecular Formula: C9H7NO4
InChi Key: WWXMVRYHLZMQIG-PLNGDYQASA-N
SMILES: OC(=O)\C=C/C1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results