accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55506-37-1

IUPAC Name: hydrogen 5-chloro-1,3-thiazol-2-amine chloride
Synonyms: hydrogen thiazole, 2-amino-5-chloro- chloride
Molecular Weight (g/mol): 171.04
Molecular Formula: C3H4Cl2N2S
InChi Key: GTMGFQYVLSQTKP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=NC=C(Cl)S1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)