accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNH3COOCH3CH3H3COOCH3CH3H3C

CAS RN 55536-71-5

IUPAC Name: 3,5-di-tert-butyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3,5-di-tert-butyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol): 309.41
Molecular Formula: C17H27NO4
InChi Key: WSWDAKKWVAVVAI-UHFFFAOYSA-N
SMILES: CC1=C(CC(C(=O)OC(C)(C)C)=C(C)N1)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)