CAS RN 55577-25-8
IUPAC Name:
2-(4-methylphenyl)-1H-indole
Synonyms:
2-(4-methylphenyl)-1H-indole
Molecular Weight (g/mol):
207.28
Molecular Formula:
C15H13N
InChi Key:
VPXGIHGJJJBJFP-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1
1
–
1
of
1
results
1
2-(p-Tolyl)indole 98.0+%, TCI America™
CAS: 55577-25-8 Molecular Formula: C15H13N Molecular Weight (g/mol): 207.28 MDL Number: MFCD00087262 InChI Key: VPXGIHGJJJBJFP-UHFFFAOYSA-N Synonym: 2-(4-Methylphenyl)indole PubChem CID: 292599 IUPAC Name: 2-(4-methylphenyl)-1H-indole SMILES: CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1