accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOCl

CAS RN 55578-23-9

IUPAC Name: N-benzyl-2-chloro-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-2-chloro-N-isopropylbenzamide
Molecular Weight (g/mol): 287.79
Molecular Formula: C17H18ClNO
InChi Key: ZNMYNGJECVXOIL-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results