accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 55589-47-4

IUPAC Name: 3-methylpyridine-2-carbaldehyde
Synonyms: 3-methylpyridine-2-carbaldehyde
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: JDYVLWWFVYNMTN-UHFFFAOYSA-N
SMILES: CC1=C(C=O)N=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)