accessibility menu, dialog, popup

UserName

Viewing profile as user:
KCH3ONSOOO

CAS RN 55589-62-3

IUPAC Name: potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
Synonyms: potassium acesulfame
Molecular Weight (g/mol): 201.24
Molecular Formula: C4H4KNO4S
InChi Key: JLEKLYQXZHJOTQ-UHFFFAOYSA-M
SMILES: [K+].CC1=CC(=O)N=S([O-])(=O)O1
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
Grade 
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)