accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2O

CAS RN 556-18-3

IUPAC Name: 4-aminobenzaldehyde
Synonyms: 4-aminobenzaldehyde
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: VATYWCRQDJIRAI-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results