accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 556-24-1

IUPAC Name: methyl 3-methylbutanoate
Synonyms: methyl isovalerate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OQAGVSWESNCJJT-UHFFFAOYSA-N
SMILES: COC(=O)CC(C)C
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (5)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)