accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHNONHOHO

CAS RN 556-33-2

IUPAC Name: 2-[2-(2-aminoacetamido)acetamido]acetic acid
Synonyms: Gly-Gly-Gly GGG triglycine diglycylglycine
Molecular Weight (g/mol): 189.17
Molecular Formula: C6H11N3O4
InChi Key: XKUKSGPZAADMRA-UHFFFAOYSA-N
SMILES: NCC(=O)NCC(=O)NCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)