accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 556-68-3

IUPAC Name: hexadecamethylcyclooctasiloxane
Synonyms: hexadecamethylcyclooctasiloxane
Molecular Weight (g/mol): 593.23
Molecular Formula: C16H48O8Si8
InChi Key: XKJMJYZFAWYREL-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results