accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 556-76-3

IUPAC Name: sodium pentyl sulfate
Synonyms: sodium pentyl sulfate
Molecular Weight (g/mol): 190.19
Molecular Formula: C5H11NaO4S
InChi Key: KLZYTGLGWPHIDV-UHFFFAOYSA-M
SMILES: [Na+].CCCCCOS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results