accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNOHNNH2

CAS RN 55621-49-3

IUPAC Name: 5,6-diamino-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5,6-diamino-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 164.17
Molecular Formula: C7H8N4O
InChi Key: BRISYWKBJNSDPL-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C2NC(=O)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results