accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOH2N

CAS RN 55695-51-7

IUPAC Name: 4-(4-chlorobenzoyl)piperidin-1-ium
Synonyms: 4-(4-chlorobenzoyl)piperidin-1-ium
Molecular Weight (g/mol): 224.71
Molecular Formula: C12H15ClNO
InChi Key: IYGWDOXHCPQXKN-UHFFFAOYSA-O
SMILES: ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results