accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnH3COOH3COO

CAS RN 557-05-1

IUPAC Name: zinc(2+) dioctadecanoate
Synonyms: zinc stearate
Molecular Weight (g/mol): 632.33
Molecular Formula: C36H70O4Zn
InChi Key: XOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES: [Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (5)
  • (3)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (7)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (7)
  • (3)
  • (8)
  • (5)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)