accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 557-11-9

IUPAC Name: (prop-2-en-1-yl)urea
Synonyms: urea, 2-propenyl-
Molecular Weight (g/mol): 100.12
Molecular Formula: C4H8N2O
InChi Key: VPJDULFXCAQHRC-UHFFFAOYSA-N
SMILES: NC(=O)NCC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results