accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OOHO(Z)

CAS RN 557-24-4

IUPAC Name: (2Z)-3-carbamoylprop-2-enoic acid
Synonyms: maleamic acid
Molecular Weight (g/mol): 115.09
Molecular Formula: C4H5NO3
InChi Key: FSQQTNAZHBEJLS-UPHRSURJSA-N
SMILES: NC(=O)\C=C/C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results