CAS RN 557-35-7
IUPAC Name:
2-bromooctane
Synonyms:
sec-octyl bromide
Molecular Weight (g/mol):
193.13
Molecular Formula:
C8H17Br
InChi Key:
FTJHYGJLHCGQHQ-UHFFFAOYNA-N
SMILES:
CCCCCCC(C)Br
1
–
1
of
1
results
1
2-Bromooctane (contains 3-Bromooctane) 85.0+%, TCI America™
CAS: 557-35-7 Molecular Formula: C8H17Br Molecular Weight (g/mol): 193.128 MDL Number: MFCD00040823 InChI Key: FTJHYGJLHCGQHQ-UHFFFAOYSA-N Synonym: 2-Octyl Bromide PubChem CID: 79046 IUPAC Name: 2-bromooctane SMILES: CCCCCCC(C)Br