accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(Z)(E)

CAS RN 557-48-2

IUPAC Name: (2E,6Z)-nona-2,6-dienal
Synonyms: (E,Z)-2,6-nonadienal trans-2,cis-6-nonadienal nonadienal nona-2,6-dienal violet leaf aldehyde nonadien-2(trans)-6-(cis)-al cucumber aldehyde trans-2-cis-6-nonadienal trans,cis-2,6-nonadienal
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: HZYHMHHBBBSGHB-ODYTWBPASA-N
SMILES: CC\C=C/CC\C=C\C=O
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)