accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 557-59-5

IUPAC Name: tetracosanoic acid
Synonyms: lignoceric acid lignozerinsaeure tetraeicosanoic acid tetraicosanoic acid tetracosansaeure
Molecular Weight (g/mol): 368.65
Molecular Formula: C24H48O2
InChi Key: QZZGJDVWLFXDLK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)