accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 557-61-9

IUPAC Name: octacosan-1-ol
Synonyms: octacosanol polycosanol montanyl alcohol cluytyl alcohol
Molecular Weight (g/mol): 410.77
Molecular Formula: C28H58O
InChi Key: CNNRPFQICPFDPO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)