accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55704-78-4

IUPAC Name: 2,5-dimethyl-1,4-dithiane-2,5-diol
Synonyms: 2,5-dimethyl-1,4-dithiane-2,5-diol
Molecular Weight (g/mol): 180.28
Molecular Formula: C6H12O2S2
InChi Key: NHKIYYMFGJBOTK-UHFFFAOYNA-N
SMILES: CC1(O)CSC(C)(O)CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results