accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH2OHN

CAS RN 55717-45-8

IUPAC Name: 6-boranylpyridin-3-ol
Synonyms: 6-boranylpyridin-3-ol
Molecular Weight (g/mol): 106.92
Molecular Formula: C5H6BNO
InChi Key: ADHMWMAEKRZWPB-UHFFFAOYSA-N
SMILES: BC1=CC=C(O)C=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)