accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55718-76-8

IUPAC Name: 2-chloro-2H-1,3,2-benzodioxaborole
Synonyms: 2-chloro-1,3,2-benzodioxaborole
Molecular Weight (g/mol): 154.36
Molecular Formula: C6H4BClO2
InChi Key: AZYGEWXDKHFOKB-UHFFFAOYSA-N
SMILES: ClB1OC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)