accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSiCH3CH3H3C

CAS RN 5573-62-6

IUPAC Name: (ethylsulfanyl)trimethylsilane
Synonyms: (ethylsulfanyl)trimethylsilane
Molecular Weight (g/mol): 134.31
Molecular Formula: C5H14SSi
InChi Key: HXAFQWICVBBXMZ-UHFFFAOYSA-N
SMILES: CCS[Si](C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)