accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3N

CAS RN 55755-16-3

IUPAC Name: 2-(2-methylphenyl)ethan-1-aminium
Synonyms: 2-methylphenethylamine
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: OWOUKRYOZIZVFK-UHFFFAOYSA-O
SMILES: CC1=CC=CC=C1CC[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results