accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNN

CAS RN 55758-02-6

IUPAC Name: 3-bromopyridine-2-carbonitrile
Synonyms: 3-bromopyridine-2-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: HCOPIUVJCIZALB-UHFFFAOYSA-N
SMILES: BrC1=CC=CN=C1C#N
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)