accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOO(E/Z)

CAS RN 5576-97-6

IUPAC Name: 1-methoxy-4-(2-nitroethenyl)benzene
Synonyms: 4-methoxy-B-nitrostyrene
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: JKQUXSHVQGBODD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C[N+]([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results