accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55779-18-5

IUPAC Name: 9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine
Synonyms: arprinocid arprinocida arprinocide arprinocidum 阿普西特 C12H9ClFN5 arpocox
Molecular Weight (g/mol): 277.69
Molecular Formula: C12H9ClFN5
InChi Key: NAPNOSFRRMHNBJ-UHFFFAOYSA-N
SMILES: NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results