accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFBrFFFFFFFF

CAS RN 558-96-3

IUPAC Name: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane
Synonyms: 1-bromoperfluorononane
Molecular Weight (g/mol): 548.97
Molecular Formula: C9BrF19
InChi Key: OWIAIPIQXHPUHV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results