accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONN

CAS RN 55804-68-7

IUPAC Name: 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carbonitrile
Synonyms: 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carbonitrile
Molecular Weight (g/mol): 266.3
Molecular Formula: C16H14N2O2
InChi Key: LGDDFMCJIHJNMK-UHFFFAOYSA-N
SMILES: O=C1OC2=C3CCCN4CCCC(C=C2C=C1C#N)=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results