accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5581-75-9

IUPAC Name: 6-phenylhexanoic acid
Synonyms: 6-phenylcaproic acid
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: JTXZPQIXIXYMDY-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)