accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55810-74-7

SH2NHCl

CAS RN 55810-74-7

IUPAC Name: 1-(1-benzothiophen-3-yl)methanamine hydrochloride
Synonyms: 1-(1-benzothiophen-3-yl)methanamine hydrochloride
Molecular Weight (g/mol): 199.7
Molecular Formula: C9H10ClNS
InChi Key: MIAPJPCDRSPNPW-UHFFFAOYSA-N
SMILES: Cl.NCC1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results