accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNHBr

CAS RN 55810-80-5

IUPAC Name: pyrrolidine hydrobromide
Synonyms: pyrrolidine hydrobromide
Molecular Weight (g/mol): 152.04
Molecular Formula: C4H10BrN
InChi Key: VFDOIPKMSSDMCV-UHFFFAOYSA-N
SMILES: Br.C1CCNC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)